n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Catalog # 8088.5000
Supplier: Avantor
CAS Number: 110-54-3
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Raumtemperatur
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
Specifications
Specification Test Results
- Assay (by GC):min. 99 %
- Aromatic Compounds (as C6H6):max. 0,02 %
- Boiling Range:max. 2°C
- Density (g/ml) at 25°C:max. 0,657
- Recorded Boiling Point:68.7°C
- Residue after Evaporation:max. 5 ppm
- Sulfur Compounds (as S):max. 0,005 %
- Thiophene:passes test
- Water (H2O):max. 0,02 %
- Water Soluble Titrable Acid (meq/g):max. 0,0003
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifications
- VE:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



