Heating tapes, HBS
Heating tape HBS, 1 m length, 250 W
Catalog # HORS020302
Supplier: HORST
Jump to:
Product Details & Documents
The heating tapes are smooth, flexible and have a multiple glass-yarn insulation as well as an additional protection braid around the heating conductors.
Specifications
- Bezeichnung:Heating tape HBS, 1 m length, 250 W
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



