Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Levetiracetam, Reference Standards | Avantor

Levetiracetam, Reference Standards

Levetiracetam Related Compound A, USP reference standard, Vial, 20 mg
Katalognr. 1359426.
Anbieter:   USP
Levetiracetam, Reference Standards
Katalognr. 1359426.
Anbieter:  USP
CAS Nummer:  
Levetiracetam, Reference Standards

Levetiracetam Related Compound A, USP reference standard, Vial, 20 mg

CAS Nummer:  
Preis
1 470.00 SFr.
Einloggen für Verfügbarkeit
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Levetiracetam Racemic Mixture: 1:1 mixture of levetiracetam S-enantiomer-(2S)-2-(2-oxopyrrolidin-1-yl)butanamide and levetiracetam R-enantiomer (2R)-2-(2-oxopyrrolidin-1-yl)butanamide

 : US Pharmacopea Reference Standard

Technische Daten

Technische Daten

  • Packung:
    Vial
  • VE:
    20 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Levetiracetam Related Compound A
  • Cat. No.:
    1359426.
  • CAS Nr.:
    102767-31-7
  • Synonym(s):
    (S)-N-(1-amino-1-oxobutan-2-yl)-4-chlorobutanamide
  • Bezeichnung:
    Levetiracetam Related Compound A
  • Formel:
    C₈H₁₅ClN₂O₂

Technische Daten