AuftragsverfolgungKontakt
Mirtazapine, Reference Standards | Avantor
Mirtazapine, Reference Standards
Mirtazapine Resolution Mixture, USP reference standard, Vial, 25 mg
Katalognr. 1444296.
Anbieter:   USP
Mirtazapine, Reference Standards
Katalognr. 1444296.
Anbieter:  USP
CAS Nummer:  
Mirtazapine, Reference Standards
Mirtazapine Resolution Mixture, USP reference standard, Vial, 25 mg
CAS Nummer:  
Preis
SFr. 1’870.00
Einloggen für Verfügbarkeit
1 * 25 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Mirtazapine Resolution Mixture: This mixture contains mirtazapine, impurity A, impurity B, impurity C, impurity D, impurity E and impurity F
Zertifizierung: US Pharmacopea Reference Standard
Technische Daten
Technische Daten
  • Packung:
    Vial
  • VE:
    25 mg
  • Normen:
    USP reference standard
  • Bezeichnung:
    Mirtazapine Resolution Mixture