Robotic tips for Eppendorf epMotion® automated pipetting system
1000 µl, -, -, 10 racks, 96 each
Catalog # 736-0763
Supplier: Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Catalog # 736-0763
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifications
- Verpackt:10 racks, 96 each
- Steril:-
- Volumen:1000 µl
- Filter:-
- Cat. No.:736-0763
- Umweltzertifizierung:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Umweltfreundlichkeit:Umweltschonende Herstellung
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..