Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Robotic tips for Eppendorf epMotion® automated pipetting system
Robotic tips for Eppendorf epMotion® automated pipetting system
1000 µl, +, +, 10 racks, 96 each
Codice catalogo 736-0761
Nome del fornitore:   Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Codice catalogo 736-0761
Nome del fornitore:  Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Robotic tips for Eppendorf epMotion® automated pipetting system
1000 µl, +, +, 10 racks, 96 each
Price
SFr. 252.00
Log in per disponibilità
1 * 960 ST
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
  • Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
  • Tips have a low percentage Coefficient of Variation (% CV)
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifiche
  • Verpackt:
    10 racks, 96 each
  • Steril:
    +
  • Volumen:
    1000 µl
  • Filter:
    +
  • Cat. No.:
    736-0761
  • Umweltzertifizierung:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Umweltfreundlichkeit:
    Umweltschonende Herstellung
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.