Robotic tips for Eppendorf epMotion® automated pipetting system
50 µl, -, -, 10 racks, 96 each
Katalognr. 736-0757
Anbieter: Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Katalognr. 736-0757
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Dokumente
Technische Daten
- Verpackt:10 racks, 96 each
- Steril:-
- Volumen:50 µl
- Filter:-
- Cat. No.:736-0757
- Umweltzertifizierung:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Umweltfreundlichkeit:Umweltschonende Herstellung
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.