Conductivity standards, potassium chloride solutions
Conductivity standard 12,88 mS/cm (KCl), 12880 µS/cm, 25 °C, 500 mL
Supplier: CONSORT
Conductivity standards, potassium chloride solutions
Catalog # CONSB561
Supplier: CONSORT
CAS Number:
Jump to:
Product Details & Documents
Potassium chloride solutions referenced at 25 °C.
- Certified to 0,5%
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- VE:500 mL
- Referenztemperatur:25 °C
- Bezeichnung:Conductivity standard 12,88 mS/cm (KCl)
- Leitfähigkeit:12880 µS/cm
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



