Test charts
Test charts with optical brightner, Schwarz/weiß kariert, A5, 148×210 mm
Katalognr. SIMEVF2346
Anbieter: Industrial Physics
Direkt zu
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
- Dimensions from A6 up to A4
- Film laminated for an excellent solvent, chemical resistance, and film spread
- Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Bestellinformationen: A certificate is available for charts with an optical brightener.
Dokumente
Technische Daten
- Format:A5
- Farbe:Schwarz/weiß kariert
- Bezeichnung:Test charts with optical brightner
- Seq:06
- B×H:148×210 mm