AuftragsverfolgungKontakt
Temperature gradient plate for MFFT test | Avantor
Temperature gradient plate for MFFT test
MFFT 20- Temperature range of −30...+250 °C with a maximum gradient on surface of 100 °C
Katalognr. SIMEVF9700
Anbieter:   Industrial Physics
Temperature gradient plate for MFFT test
Katalognr. SIMEVF9700
Anbieter:  Industrial Physics
Temperature gradient plate for MFFT test
Temperature gradient plate for MFFT test
MFFT 20- Temperature range of −30...+250 °C with a maximum gradient on surface of 100 °C
Preis
46 060.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Temperature gradient plate for determining minimum film forming temperature (MFFT) test. The instrument can be used to define the white point/glass transition temperature of dispersion materials, synthetic resins, enamels and the blocking power and stacking capability of coated papers, foils, prints.
  • Ground hard-chrome plated metal platen as to deposit the specimen
  • Displays nominal and actual value of hot and cold side
  • Electronic controller with digital display, temperature preselecting and overshoot protection
  • Integrated membrane drying unit with flow meter
The temperature is controlled through an integrated digital controller with a digital display and measuring-point selector that covers the whole range. To determine the blocking power the specimens on the platen are weighed by defined loads to simulate the stacking capability.
MFFT 10: Includes 10 built-in temperature sensors and hinged transparent cover made of PMMA. Recommended standard cryostat type is C10K20.
MFFT 20: Includes 20 built-in temperature sensors and hinged transparent cover made of stainless steel and safety glass for temperatures up to +250 °C. Recommended standard cryostat type is KWK10.
W×D×H (with closed cover): 350×800×320 mm
Zertifizierung: Conforms to DIN ISO 2115 – DIN 53366 – ASTM D 2354 – ASTM D 1465 – ISO 2115 – ISO/DIS 4622.
Bestellinformationen: Optional cooling unit must be ordered separately.
Dokumente
Technische Daten
  • Bezeichnung:
    MFFT 20- Temperature range of −30...+250 °C with a maximum gradient on surface of 100 °C
  • Seq:
    02