AuftragsverfolgungKontakt
Temperature indicating strips
Temperature indicating strips
77…116 °C
Codice catalogo SIMETC8020
Nome del fornitore:   Industrial Physics
Temperature indicating strips
Codice catalogo SIMETC8020
Nome del fornitore:  Industrial Physics
Temperature indicating strips
Temperature indicating strips
77…116 °C
Price
37.20 SFr.
Log in per disponibilità
1 * 10 ST
Product Details & Documents
These temperature indicating strips are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construction businesses. They are particularly useful for indicating temperatures of moving parts.
  • Self adhesive foils to measure maximum surface temperatures
  • Indicators change colour when marked temperature is reached and the change is irreversible
  • Highest temperature remains visible for months
Informazioni per l'ordine: Series 5 and Series 8 contains 10 stickers per sheet.
Specifiche
  • Seq:
    09
  • Messbereich:
    77…116 °C