5-Bromphthalid ≥98%
Glass bottle for solids, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: 5-Bromoisobenzofuran-1(3H)-one
- CAS Nummer:64169-34-2
- Formel:C₈H₅BrO₂
- MDL-Nummer:MFCD01797360
- Molekulargewicht:213.03 g/mol
- Schmelzender Platingehalt:162…166 °C
- Lagertemperatur:Raumtemperatur
- EINECS:447-190-5
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFAH33891
Sicherheitsdatenblatt (SDS)
Organic Building Blocks and Intermediates
Anwendungshinweise
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



