Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
5-Bromphthalid ≥98%

5-Bromphthalid ≥98%

Glass bottle for solids, 25 g
Catalog # H33891.14
Irritant
5-Bromphthalid ≥98%
Catalog # H33891.14
CAS Number:  64169-34-2
5-Bromphthalid ≥98%
5-Bromphthalid ≥98%

Glass bottle for solids, 25 g

CAS Number:  64169-34-2
Price
SFr. 166.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  5-Bromoisobenzofuran-1(3H)-one
  • CAS Number:64169-34-2
  • Formula:C₈H₅BrO₂
  • MDL Number:MFCD01797360
  • Molecular Weight:213.03 g/mol
  • Melting Pt:162…166 °C
  • Storage Temperature:Raumtemperatur
  • EINECS:447-190-5
Specifications

Specifications

  • Packung:
    Glass bottle for solids
  • VE:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.