AuftragsverfolgungKontakt
Manual detergent, Emerald
Manual detergent, Emerald
Emerald neutral detergent for manual cleaning, 1x 1000 litre IBC
Catalog # TRSTCRY310
Supplier:   TRISTEL SOLUTIONS
Manual detergent, Emerald
Catalog # TRSTCRY310
Supplier:  TRISTEL SOLUTIONS
Manual detergent, Emerald
Manual detergent, Emerald
Emerald neutral detergent for manual cleaning, 1x 1000 litre IBC
Price
Inquire For Price
Log in for availability
1 * 1.000 L
Product Details & Documents
A high foam, neutral, detergent for manual cleaning.
  • Neutral detergent for light to medium soiling
  • Dilution for use: Between 0,5% to 2% v/v
A manual detergent for the removal of light to medium soiling. Areas of use include walls, floors, surfaces, ceilings, equipment and utensils. Emerald is a soft metal safe detergent suitable for use on a variety of surfaces including plastics, silicone, vinyl and aluminium.
The crystel detergents are specifically formulated for the removal of the various residues that are present in the manufacturing and production process within pharmaceutical and personal care industries.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Delivery information: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Packaging: Available in 5, 20, 200 and 1000 litre pack sizes.
Documents
Specifications
  • Bezeichnung:
    Emerald neutral detergent for manual cleaning
  • Seq:
    04
  • Verpackt:
    1x 1000 litre IBC