Biotin labelling kit
Biotin Labeling Kit with amine (-NH2) reactive group, 1 KIT
Katalognr. ABNOKA0003
Anbieter: Abnova
Direkt zu
Product Details & Documents
Biotin Labelling Kit is used for simple and rapid preparation of biotin-labeled antibodies and proteins.
Achtung! Bitte beachten: Store at 4 °C. For research use only.
Dokumente
Literature reference - PubMed ID 23022042 Literature reference - PubMed ID 24814701 Literature reference - PubMed ID 26119087
Weitere Produktdetails
sds_ABNOKA0003 sds_ABNOKA0004
Technisches Datenblatt
Technische Daten
Technische Daten
- VE:1 KIT
- Bezeichnung:Biotin Labeling Kit with amine (-NH<sub>2</sub>) reactive group