Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Biotin labelling kit | Avantor
Biotin labelling kit
Biotin Labeling Kit with amine (-NH2) reactive group, 1 KIT
Katalognr. ABNOKA0003
Anbieter:   Abnova
Biotin labelling kit
Katalognr. ABNOKA0003
Anbieter:  Abnova
CAS Nummer:  
Biotin labelling kit
Biotin labelling kit
Biotin Labeling Kit with amine (-NH2) reactive group, 1 KIT
CAS Nummer:  
Preis
SFr. 403.00
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Biotin Labelling Kit is used for simple and rapid preparation of biotin-labeled antibodies and proteins.
Achtung! Bitte beachten: Store at 4 °C. For research use only.
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    Biotin Labeling Kit with amine (-NH<sub>2</sub>) reactive group
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.