Diclofenac-Natrium
Glasflasche für Feststoffe, 50 g
Catalog # CAYM70680-50
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt, GP-45840, [o-(2,6-dichloroanilino) phenyl]acetic acid sodium salt, 2-[(2,6-Dichlorophenyl)amino]-benzeneacetic acid monosodium salt, Sodium [o-[(2,6-dichlorophenyl) amino] phenyl] acetate
- CAS Number:15307-79-6
- Formula:C₁₄H₁₀Cl₂NNaO₂
- MDL Number:MFCD00082251
- Molecular Weight:318.1 g/mol
- EINECS:239-346-4
- UN:2811
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



