PPM-18 ≥95%
Glass vial, 25 mg
Katalognr. CAYM13327-25
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
- Inhibits activation of NF-κB
An inhibitor NF-κB activation in vitro and in vivo (IC₅₀= 5 µm).
Synonyme: NSC 73233
- CAS Nummer:65240-86-0
- Lagertemperatur:Gefrierschrank
Technische Daten
Ergebnisse des Spezifikationstests
- Formulation:A crystalline solid
- Purity:≥95%
- Stability:≥2 years
Technische Daten
- Packung:Glass vial
- VE:25 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



