AuftragsverfolgungKontakt
(±)-7-HDoHE
250 µG
Catalog # CAYM33300-250
Supplier:   Cayman Chemical
(±)-7-HDoHE
Catalog # CAYM33300-250
Supplier:  Cayman Chemical
CAS Number:  90780-55-5
(±)-7-HDoHE
250 µG
CAS Number:  90780-55-5
Price
SFr. 1’180.00
Log in for availability
1 * 250 µG
Product Details & Documents
  • CAS Number:90780-55-5
  • Formula:C₂₂H₃₂O₃
  • UN:1170
  • ADR:3,II
Specifications
Specifications
  • VE:
    250 µG
Specifications