(±)-7-HDoHE
100 µG
Codice catalogo CAYM33300-100
Nome del fornitore: Cayman Chemical
Vai a
Product Details & Documents
- Numero CAS:90780-55-5
- Formula:C₂₂H₃₂O₃
- UN:1170
- ADR:3,II
Specifiche
Specifiche
- VE:100 µG
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



