Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Methylpalmitat

Methylpalmitat

Glasflasche für Feststoffe, 10 g
Catalog # CAYM10007358-10
Supplier:   Cayman Chemical
Methylpalmitat
Catalog # CAYM10007358-10
Supplier:  Cayman Chemical
CAS Number:  112-39-0
Methylpalmitat

Glasflasche für Feststoffe, 10 g

CAS Number:  112-39-0
Price
SFr. 169.00
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
  • CAS Number:112-39-0
  • Formula:C₁₇H₃₄O₂
  • MDL Number:MFCD00008994
  • Molecular Weight:270.46 g/mol
  • Boiling Pt:417 °C (1013 hPa)
  • Melting Pt:29,9 °C
  • Density:0,8804 g/cm³ (20 °C)
  • EINECS:203-966-3

Documents

Specifications

Specifications

  • Packung:
    Glasflasche für Feststoffe
  • VE:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.