3-Methoxythiobenzamid
Glass bottle for solids, 1 g
Katalognr. CAYM10553-1
Anbieter: Cayman Chemical
Direkt zu
Product Details & Documents
Synonyme: 3-Methoxybenzothioamide, 3-Methoxybenzenecarbothioamide
- CAS Nummer:64559-06-4
- Formel:C₈H₉NOS
- MDL-Nummer:MFCD04627361
- Molekulargewicht:167.23 g/mol
Technische Daten
Technische Daten
- Packung:Glass bottle for solids
- VE:1 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



