Thermal transfer printer labels, type B-424
Printer labels, B-424, THT-25-424-1, Papier, Permanent, gummibasiert, Thermal transfer, Weiß, 360X-Plus II, 101×152 mm, 1000 labels/roll
Catalog # BRDY030153
Supplier: Brady
Jump to:
Product Details & Documents
Printable, paper labels for thermal transfer printers.
- Good contrast and smear resistance
Labels are ideally suited for use on smooth surfaces.
Specifications
- Druckermodell:360X-Plus II
- Printing method:Thermal transfer
- Typ:B-424
- Farbe:Weiß
- Bezeichnung:Printer labels
- Abmessungen:101×152 mm
- Material:Papier
- Seq:13
- Modell:THT-25-424-1
- Verpackt:1000 labels/roll
- Klebstoff:Permanent, gummibasiert