Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Thermal transfer printer labels, type B-424
Thermal transfer printer labels, type B-424
Printer labels, B-424, THT-25-424-1, Papier, Permanent, gummibasiert, Thermal transfer, Weiß, 360X-Plus II, 101×152 mm, 1000 labels/roll
Catalog # BRDY030153
Supplier:   Brady
Thermal transfer printer labels, type B-424
Catalog # BRDY030153
Supplier:  Brady
Thermal transfer printer labels, type B-424
Thermal transfer printer labels, type B-424
Printer labels, B-424, THT-25-424-1, Papier, Permanent, gummibasiert, Thermal transfer, Weiß, 360X-Plus II, 101×152 mm, 1000 labels/roll
Price
396.00 SFr.
Log in for availability
1 * 1 ROU
Product Details & Documents
Printable, paper labels for thermal transfer printers.
  • Good contrast and smear resistance
Labels are ideally suited for use on smooth surfaces.
Specifications
  • Druckermodell:
    360X-Plus II
  • Printing method:
    Thermal transfer
  • Typ:
    B-424
  • Farbe:
    Weiß
  • Bezeichnung:
    Printer labels
  • Abmessungen:
    101×152 mm
  • Material:
    Papier
  • Seq:
    13
  • Modell:
    THT-25-424-1
  • Verpackt:
    1000 labels/roll
  • Klebstoff:
    Permanent, gummibasiert
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.