Microplates, 96-well, non binding
96-well microplate, F-bottom, non binding, chimney well, μClear® bottom, Weiß/klar
Katalognr. 738-0171
Anbieter: Greiner Bio-One
Direkt zu
Product Details & Documents
Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity by reducing background and improving signal-to-noise ratio.
Technische Daten
- Farbe:Weiß/klar
- Bezeichnung:96-well microplate, F-bottom, non binding, chimney well, μClear® bottom
- Seq:06
- Cat. No.:738-0171
- Umweltzertifizierung:ISO 14001 (241002); ISO 50001 (241002)
- Umweltfreundlichkeit:Umweltschonende Herstellung