Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Water purification systems, AFS® 8/16/24

Water purification systems, AFS® 8/16/24

AFS® 16 water purification system (EU plug, manual in Italian)
Catalog # MILLZAFS0160IT
Water purification systems, AFS® 8/16/24
Catalog # MILLZAFS0160IT
Supplier:  
Water purification systems, AFS® 8/16/24

AFS® 16 water purification system (EU plug, manual in Italian)

Discontinued
Product Details & Documents

Economical water system, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 l/min. For clinical analysers with pure water needs up to 480 L daily

  • Cost-effective: Optimised lifetimes for pretreatment and polishing packs, low feed water use
  • Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
  • Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
  • Intuitive icon-based system display facilitates maintenance
  • Ergonomic pack locking system makes pack changes easy

Complementary water purification techniques, including complete pretreatment, advanced reverse osmosis (RO), exposure to a bactericidal UV lamp, and final 0,22 μm filtration, provide consistent water quality meeting CLRW standards.

Specifications
  • Bezeichnung:
    AFS® 16 water purification system (EU plug, manual in Italian)
  • Seq:
    15
  • Produktionsfließrate:
    16 L/h @ 25 °C
  • Cat. No.:
    MILLZAFS0160IT
  • TOC-Gehalt:
    <30 ppb
  • Modell:
    AFS® 16
  • Bakterien:
    < 1 CFU/ml
  • B×T×H:
    268×426×585 mm
  • Speisewasser:
    Portable tap water at a maximum pressure of 6 bar
  • Spezifischer Widerstand bei +25 °C:
    >15 MΏ.cm
  • Gewicht:
    <28 kg
Accessories

Product Information

  • Catalog #
    Bezeichnung
    Availability
    Price
  • MILLTANK00200
    Storage tank (no relay), 200 l
    Log in for availability
    1 * 1 ST - SFr. 8’580.00

Product Information

  • Catalog #
    Bezeichnung
    Availability
    Price
  • MILLCP2ALLREST
    Q-Gard® A polishing cartridge with virgin ion-exchange resins
    Log in for availability
    1 * 1 ST - SFr. 678.00

Product Information

  • Catalog #
    Bezeichnung
    Availability
    Price
  • MILLPR0G00003
    Progard® 3 Pretreatment Pack
    Log in for availability
    1 * 1 ST - SFr. 621.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.