Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Water purification systems, AFS® 8/16/24

Water purification systems, AFS® 8/16/24

AFS® 24 water purification system (EU plug, manual in Spanish)
Katalognr. MILLZAFS0240SP
Water purification systems, AFS® 8/16/24
Katalognr. MILLZAFS0240SP
Anbieter:  
Water purification systems, AFS® 8/16/24

AFS® 24 water purification system (EU plug, manual in Spanish)

Artikel gestrichen
Product Details & Documents

Economical water system, producing CLRW (Clinical Laboratory Reagent Water) quality with a dispensing flow rate to analyser up to 2 l/min. For clinical analysers with pure water needs up to 480 L daily

  • Cost-effective: Optimised lifetimes for pretreatment and polishing packs, low feed water use
  • Full monitoring and automatic water quality archiving; information can be stored for up to 6 months
  • Small footprint, allowing installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
  • Intuitive icon-based system display facilitates maintenance
  • Ergonomic pack locking system makes pack changes easy

Complementary water purification techniques, including complete pretreatment, advanced reverse osmosis (RO), exposure to a bactericidal UV lamp, and final 0,22 μm filtration, provide consistent water quality meeting CLRW standards.

Technische Daten
  • Bezeichnung:
    AFS® 24 water purification system (EU plug, manual in Spanish)
  • Seq:
    25
  • Produktionsfließrate:
    24 L/h @ 25 °C
  • Cat. No.:
    MILLZAFS0240SP
  • TOC-Gehalt:
    <30 ppb
  • Modell:
    AFS® 24
  • Bakterien:
    < 1 CFU/ml
  • B×T×H:
    268×426×585 mm
  • Speisewasser:
    Portable tap water at a maximum pressure of 6 bar
  • Spezifischer Widerstand bei +25 °C:
    >15 MΏ.cm
  • Gewicht:
    <28 kg
Accessories

Produktinformationen

  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • MILLTANK00200
    Storage tank (no relay), 200 l
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 8’580.00

Produktinformationen

  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • MILLCP2ALLREST
    Q-Gard® A polishing cartridge with virgin ion-exchange resins
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 678.00

Produktinformationen

  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • MILLPR0G00003
    Progard® 3 Pretreatment Pack
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 621.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.