Dichtungen
Rotor seal, SIL-10ADvp, 228-21217-97, SHIM
Catalog # CTSACTS-10942
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Bezeichnung:Rotor seal
- Seq:51
- Erstausrüster:SHIM
- Erstausrüster-Äquivalent:228-21217-97
- Modell:SIL-10ADvp