Dichtungen
Plunger rinse seal, LC-10Ai, LC-10AS, LC-10AT, LC-10ATvp, 228-28499-00, SHIM
Katalognr. CTSACTS-10583
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Plunger rinse seal
- Seq:29
- Erstausrüster:SHIM
- Erstausrüster-Äquivalent:228-28499-00
- Modell:LC-10Ai, LC-10AS, LC-10AT, LC-10ATvp
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.