Dichtungen
HPMV Dichtung, gold, 717, 715, 2690, 2690D, 2695, 2695D, WAT045454, WATE
Catalog # 560-0079
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Bezeichnung:HPMV Dichtung, gold
- Seq:11
- Erstausrüster:WATE
- Erstausrüster-Äquivalent:WAT045454
- Modell:717, 715, 2690, 2690D, 2695, 2695D