AuftragsverfolgungKontakt
Dichtungen
Kopf-Kolbendichtung, ACQUITY H-Class QSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM, 700002599, WATE
Codice catalogo 560-0105
Nome del fornitore:   SCIENCIX
Dichtungen
Codice catalogo 560-0105
Nome del fornitore:  SCIENCIX
Dichtungen
Dichtungen
Kopf-Kolbendichtung, ACQUITY H-Class QSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM, 700002599, WATE
Price
SFr. 152.00
Log in per disponibilità
1 * 2 ST
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifiche
  • Bezeichnung:
    Kopf-Kolbendichtung
  • Seq:
    09
  • Erstausrüster:
    WATE
  • Erstausrüster-Äquivalent:
    700002599
  • Modell:
    ACQUITY H-Class QSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM