Dichtungen
Kopf-Kolbendichtung, ACQUITY H-Class QSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM, 700002599, WATE
Codice catalogo 560-0105
Nome del fornitore: SCIENCIX
Vai a
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifiche
- Bezeichnung:Kopf-Kolbendichtung
- Seq:09
- Erstausrüster:WATE
- Erstausrüster-Äquivalent:700002599
- Modell:ACQUITY H-Class QSM, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, nanoACQUITY UPLC BSM/ASM