Dichtungen
Plunger seal, black, Constametric I, II, III, CM3000, CM3200, CM3500, CM4000, CM4100, 206129001, RUNC
Katalognr. CTSACTS-20612
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Plunger seal, black
- Erstausrüster:RUNC
- Erstausrüster-Äquivalent:206129001
- Modell:Constametric I, II, III, CM3000, CM3200, CM3500, CM4000, CM4100