Dichtungen
Plunger seal, gold, Constametric I, II, III, CM3000, CM3200, CM3500, CM4000, CM4100, 206156001, RUNC
Katalognr. CTSACTS-2061
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Plunger seal, gold
- Seq:42
- Erstausrüster:RUNC
- Erstausrüster-Äquivalent:206156001
- Modell:Constametric I, II, III, CM3000, CM3200, CM3500, CM4000, CM4100