Dichtungen
Plunger seal, black, 114M, 116, 118, 125, 126, 127, 128, 237162, 728770, Beckman
Katalognr. CTSACTS-7162
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Plunger seal, black
- Erstausrüster:Beckman
- Erstausrüster-Äquivalent:237162, 728770
- Modell:114M, 116, 118, 125, 126, 127, 128