Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Glass Microfiber Filter Paper | Avantor

Glass Microfiber Filter Paper

GF/A, 19 mm
Katalognr. PRATA0908837
Anbieter:   PRAT DUMAS
Glass Microfiber Filter Paper
Katalognr. PRATA0908837
Anbieter:  PRAT DUMAS
Glass Microfiber Filter Paper
Glass Microfiber Filter Paper

GF/A, 19 mm

Preis
SFr. 107.00
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents

These glass microfiber filter papers allow fast filtration and high particle retention. They can be used at temperatures up to 380 °C.

  • Chemical compatibility with many solvents and with reagents
  • Suitable for microfiltration
Technische Daten
  • Sorte:
    GF/A
  • Seq:
    1
  • Größe:
    19 mm
  • Cat. No.:
    PRATA0908837
  • Stärke:
    190 µm
  • Gewicht:
    37 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.