AuftragsverfolgungKontakt
Splash heads, with sinter, Quickfit® | Avantor
Splash heads, with sinter, Quickfit®
29/32 NS, 29/32 NS
Katalognr. 201-0576
Anbieter:   SciLabware
Splash heads, with sinter, Quickfit®
Katalognr. 201-0576
Anbieter:  SciLabware
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
29/32 NS, 29/32 NS
Preis
SFr. 234.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
PYREX® borosilicate glas
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.
Technische Daten
  • Kern:
    29/32 NS
  • Hülse:
    29/32 NS
  • Seq:
    03