Veterinary needles
Veterinary Luer needles, 12, 25 mm
Katalognr. POPR7230
Anbieter: CADENCE SCIENCE
Direkt zu
Product Details & Documents
These hypodermic needles are all fitted with a large knurled nickel plated hub which allows for secure gripping even under the most difficult circumstances.
Technische Daten
- Gauge:12
- Bezeichnung:Veterinary Luer needles
- Länge:25 mm
- Seq:02