Veterinary needles
Veterinary Luer needles, 15, 38 mm
Codice catalogo POPR7268
Nome del fornitore: CADENCE SCIENCE
Vai a
Product Details & Documents
These hypodermic needles are all fitted with a large knurled nickel plated hub which allows for secure gripping even under the most difficult circumstances.
Specifiche
- Gauge:15
- Bezeichnung:Veterinary Luer needles
- Länge:38 mm
- Seq:06