Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Carbohydrate coupling resin
Carbohydrate coupling resin
Carbohydrate Immobilisation Kit, for 5×2 ml columns, 1 KIT
Katalognr. 786-807
Anbieter:   G-Biosciences
Carbohydrate coupling resin
Katalognr. 786-807
Anbieter:  G-Biosciences
CAS Nummer:  
Carbohydrate coupling resin
Carbohydrate coupling resin
Carbohydrate Immobilisation Kit, for 5×2 ml columns, 1 KIT
CAS Nummer:  
Preis
780.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 KIT
Product Details & Documents
Carbohydrate coupling resin is designed for the covalent immobilisation of carbohydrate containing molecules, including glycoproteins, to agarose beads. It is ideal for the coupling of polyclonal antibodies as it allows for the optimal orientation of the antibodies for affinity purification.
The Carbohydrate Immobilization Kit is supplied with carbohydrate coupling resin columns for the preparation of 2 ml affinity columns, a coupling catalyst, SpinOUT™ GT-600 Desalting Columns for removing oxidising reagents, the oxidizing reagent (sodium meta-periodate) and relevant buffers.
Technische Daten
Technische Daten
  • VE:
    1 KIT
  • Bezeichnung:
    Carbohydrate Immobilisation Kit, for 5×2 ml columns
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.