Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Carbohydrate coupling resin

Carbohydrate coupling resin

Carbohydrate coupling resin, 10 mL
Catalog # 786-808
Supplier:   G-Biosciences
Carbohydrate coupling resin
Catalog # 786-808
Supplier:  G-Biosciences
CAS Number:  
Carbohydrate coupling resin
Carbohydrate coupling resin

Carbohydrate coupling resin, 10 mL

CAS Number:  
Price
SFr. 223.00
Log in for availability
1 * 10 mL
Product Details & Documents

Carbohydrate coupling resin is designed for the covalent immobilisation of carbohydrate containing molecules, including glycoproteins, to agarose beads. It is ideal for the coupling of polyclonal antibodies as it allows for the optimal orientation of the antibodies for affinity purification.

The Carbohydrate Immobilization Kit is supplied with carbohydrate coupling resin columns for the preparation of 2 ml affinity columns, a coupling catalyst, SpinOUT™ GT-600 Desalting Columns for removing oxidising reagents, the oxidizing reagent (sodium meta-periodate) and relevant buffers.

Specifications

Specifications

  • VE:
    10 mL
  • Bezeichnung:
    Carbohydrate coupling resin

Specifications