Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Transport container with screw cap

60 ml, 30 mm, 126 mm, Multi-purpose, 300/300 (bag/ctn)
Katalognr. BOET08.023.0000
Anbieter:   BOETTGER PAUL
Transport container with screw cap
Katalognr. BOET08.023.0000
Anbieter:  BOETTGER PAUL
Transport container with screw cap
Transport container with screw cap

60 ml, 30 mm, 126 mm, Multi-purpose, 300/300 (bag/ctn)

Preis
SFr. 96.00
Einloggen für Verfügbarkeit
1 * 300 ST
Product Details & Documents

Made of PP, translucent. Mailing containers for medical and biohazard samples.

  • Optimal protection from external elements through cylindrical design
  • Absorbent paper insert provides additional protection, easy to change, multiple-use very simple
  • Biohazard and recycling symbol clearly visible

Mailing containers Ø 30 mm fits into current mailboxes with 35 mm-wide slot, containers 44/114 mm suitable for mailing common culture mediums and containers 30/126 mm with cap suitable for Port-a-Cul.

 : Corresponding to DIN EN 829 (with insert/print).

 : Transport tubes and screw caps have to be ordered separate.

 : *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904

Technische Daten
  • Ext.Ø:
    30 mm
  • Verpackt:
    300/300 (bag/ctn)
  • Ausführung:
    Multi-purpose
  • Kapazität:
    60 ml
  • Höhe:
    126 mm
  • Seq:
    04