Kryoboxen, Karton
5×5, 24 mm
HERA820025
Cryo resistant cardboard.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
- Fächer:5×5
- Für Röhrchen Ø:24 mm
- Seq:06