Kryoboxen, Karton
8×8, 14 mm
Katalognr. HERA5960
Anbieter: Thermo Scientific
Direkt zu
Product Details & Documents
Cryo resistant cardboard.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
Dokumente
HERA820002_TDS_EN
Technisches Datenblatt
Technische Daten
- Fächer:8×8
- Für Röhrchen Ø:14 mm
- Seq:08