Kryoboxen, Karton
7×7, 17 mm
HERA820049
Cryo resistant cardboard.
- Use with cardboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7×7 , 8×8, 9×9 or 10×10 grids
For storage of cryovials at low temperatures.
- Fächer:7×7
- Für Röhrchen Ø:17 mm
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.