Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Zubehör | Avantor
Zubehör
PTFE replacement tubing, 500 mm; with a long screw connection; Ext.Ø 1,6 mm, Int.Ø 1,0 mm; with OMNIFIT® gripper
Katalognr. HELL040-202-72
Anbieter:   Hellma
Zubehör
Katalognr. HELL040-202-72
Anbieter:  Hellma
Zubehör
Zubehör
PTFE replacement tubing, 500 mm; with a long screw connection; Ext.Ø 1,6 mm, Int.Ø 1,0 mm; with OMNIFIT® gripper
Preis
SFr. 75.30
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Technische Daten
  • Bezeichnung:
    PTFE replacement tubing, 500 mm; with a long screw connection; Ext.Ø 1,6 mm, Int.Ø 1,0 mm; with OMNIFIT® gripper
  • Seq:
    6
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.