Zubehör
PTFE replacement tubing, 500 mm; with a long screw connection; Ext.Ø 1,6 mm, Int.Ø 1,0 mm; with OMNIFIT® gripper
Katalognr. HELL040-202-72
Anbieter: Hellma
Direkt zu
Product Details & Documents
Technische Daten
- Bezeichnung:PTFE replacement tubing, 500 mm; with a long screw connection; Ext.Ø 1,6 mm, Int.Ø 1,0 mm; with OMNIFIT® gripper
- Seq:6