Spanner wrenches
Schraubenschlüssel, für Nüsse Ø 95 - 100 mm, für HiScale™ 50 Säulen, Verzinkter Stahl
Catalog # 28-9647-78
Supplier: Cytiva
Jump to:
Product Details & Documents
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.
Specifications
- Bezeichnung:Schraubenschlüssel, für Nüsse Ø 95 - 100 mm, für HiScale™ 50 Säulen
- Material:Verzinkter Stahl
- Seq:03