Orion Star™ A326 Multi-Parameter Meter (pH/RDO/DO), Handheld
Product Details & Documents
This rugged, waterproof portable meter is designed for a wide range of pH, mV, ORP, dissolved oxygen and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP67-rated housing. The meter offers two measuring channels allowing users to measure pH and dissolved oxygen simultaneously or view each channel separately.
- Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups
- Automatically compensate dissolved oxygen readings using ATC temperature, manual salinity input and built-in barometer for barometric pressure readings
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
Measure pH, mV, relative mV or ORP with temperature on channel one and measure dissolved oxygen in percent saturation or mg/L with temperature using RDO optical DO probes or polarographic DO probes on channel two. Warrenty; 3 years.
: CE, TUV 3-1, FCC Class A.
: Supplied with four AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8107UWMMD ROSS gel-filled epoxy pH/ATC Triode (3 m cable), 087010MD RDO optical DO probe (3 m cable), pH 4,01/pH 7,00/pH 10,01 buffer pouches, Orion ROSS electrode storage solution, Orion 1413 µS conductivity standard pouches, rinse solution pouches, protective armor with electrode holder and hard carrying case.
Technische Daten
- Bezeichnung:Handheld pH/DO meter Orion Star A326, Kit incl. 8107UWMMD ROSS Triode 3-in-1 pH/ATC probe, 087010MD RDO probe with calibration sleeve, cup, 3×10 buffer pouches, storage and rinsing solutions, protective armor with electrode holder, hard carrying case
- Seq:02
- Kalibrierpunkte:1 point relative mV (RmV) mode, 1 point ORP mode
- DO Genauigkeit:Polarographic: ±0,2 mg/l or ±2% of reading, whichever is greater
RDO: ±0,1 mg/l (up to 8 mg/l), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l) - Sauerstoff - Sättigung:0,0 - 600,0 % (polarographic); 0,0 - 500,0 % (RDO)
- Anzahl der Kanäle:2
- DO Konzentration:0 - 90 mg/L (polarographic); 0 - 50 mg/L (RDO)
- Pressure accuracy:6 mm Hg
- Temperatur - Kompensation:Automatic or manual
- Pressure resolution:1 mm Hg
- mV - Bereich:±2000,0
- DO Kalibrierung:Water-saturated air, air saturated water, manual (Winkler) and zero point
- mV - Genauigkeit:±0,2 mV or 0,05% of reading whichever is greater
- Akku-Typ:4 AA (included)
- Datenprotokollierung - Logfunktion:Manual / Automatic with AUTO-READ™
- Sauerstoff - Auflösung:0,1 / 0,01 mg/L
- Ausgänge:RS232, USB
- Temperaturgenauigkeit:±0,1 °C
- Temperaturauflösung:0,1 °C, 0,1 °F
- Konformität:IP 67
- Temperatur-Kalibrierung:1 point temperature offset calibration
- Gewicht:0,5 kg
- pH - Genauigkeit:±0,002
- pH - Bereich:–2,000...+20,000
- Kalibrierung:pH with calibration editing option, relative mV (RmV), ORP, dissolved oxygen and temperature
- Sauerstoff – Luftdruckbereich:Automatic or manual, 450 - 850 mmHg
- Cat. No.:665-0280
- Anschlüsse:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 9 pin MiniDIN (RDO optical or polarographic DO sensor with built-in temperature)
- Modell:A326
- Sprachen:English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
- Schutzklasse:IP 67
- Kapazität Datenprotokollierung:5000 with date and time stamp
- B×T×H:240×100×65 mm
- pH - Auflösung:0,1 / 0,01 / 0,001
- DO resolution:0.1, 1 % saturation
- Alarm:High/low limit alarm, calibration due alarm
- mV - Auflösung:0,1
- Speicher:Non-volatile memory preserves data log, calibration log and meter settings
- Batterielebensdauer:800 hours
- Stromversorgung:AC adapter or 4 AA batteries
- pH - Kalibrierung:Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- Temperaturbereich:–5,0...+105,0 °C
- Display:Graphic LCD with backlight
Accessories
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- 662-1140Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cableThermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus metersEinloggen für Verfügbarkeit1 * 1 SET - SFr. 94.40
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- 662-1547Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 89.50
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-ARProtective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holderEinloggen für Verfügbarkeit1 * 1 ST - SFr. 126.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-ESCDOrion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeveEinloggen für Verfügbarkeit1 * 1 ST - SFr. 36.50
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-ESPHOrion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeveEinloggen für Verfügbarkeit1 * 1 ST - SFr. 36.50
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-CSHard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeveEinloggen für Verfügbarkeit1 * 1 ST - SFr. 178.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIO1210005Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the fieldEinloggen für Verfügbarkeit1 * 1 ST - SFr. 199.00
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- ORIO1010005Cable adapter, for connecting the RS232 port of a meter to a USB portOrion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 162.00
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- ORIO1010053RS232 cable, for connecting a meter to a PC or printerOrion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 56.10
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



