Multi-parameter meter (pH/mV/conductivity), handheld, Orion Star™ A325
Product Details & Documents
This rugged, waterproof portable meter is designed for a wide range of pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity and temperature testing and field applications. It can be used in the most demanding locations thanks to its IP67-rated housing. The meter offers two measuring channels allowing users to measure pH and conductivity simultaneously or view each channel separately.
- Up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously
- Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
Measure pH, mV, relative mV or ORP with temperature on channel one and measure conductivity, TDS, salinity or resistivity with temperature on channel two.
Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate. Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures.
Lieferinformationen: Supplied with four with 4 AA batteries, literature CD, printed quick start guide, computer interface cable and meter test certificate.
The kit additionally includes: 8107UWMMD ROSS Ultra Triode gel-filled epoxy-body pH/ATC electrode (3 m cable), 013010MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor (3 m cable), Orion pH 4/7/10 buffer pouches (10 packs each), ROSS electrode storage solution (475 ml), Orion rinse solution pouches (10 packs), Orion 1413 µS conductivity standard pouches (10 packs), Orion Star A series portable meter armor and Orion Star A series hard-sided field case.
Technische Daten
- Bezeichnung:Handheld pH/conductivity meter Orion Star A325, instrument alone
- Seq:01
- Kalibrierpunkte:1 point relative mV (RmV) mode, 1 point ORP mode
- Auflösung der Leitfähigkeit:0,001 μS minimum, auto ranging, up to 4 significant digits
- Eingang:Two measuring channels
- Anzahl der Kanäle:2
- TDS - Umrechnungsfaktor:Linear (0,02 - 9,99)
- TDS - Auflösung:0,001 ppm minimum, auto ranging, up to 4 significant digits
- Salinität - Genauigkeit:0,5% of reading ±1 digit
- Leitfähigkeitsbereich:0,001 µS/cm - 3000 mS/cm
- TDS - Bereich:0,001 - 200,0 ppm
- Temperatur - Kompensation:Automatic or manual
- mV - Bereich:±2000,0
- Spezifischer Widerstand - Bereich:2 Ω/cm - 100 MΩ/cm
- mV - Genauigkeit:±0,2 mV or ±0,05% of reading, whichever is greater
- Akku-Typ:4 AA (included)
- Datenprotokollierung - Logfunktion:Manual / Automatic with AUTO-READ™
- Spezifischer Widerstand - Genauigkeit:0,5% of reading ±1 digit
- Ausgänge:RS232, USB
- Temperaturgenauigkeit:±0,1 °C
- Temperaturauflösung:0,1 °C
- Konformität:IP 67
- Gewicht:0,5 kg
- pH - Genauigkeit:±0,002
- pH - Bereich:–2,000...+20,000
- Salinität - Auflösung:0,01 psu or 0,01 ppt minimum, auto ranging
- TDS - Genauigkeit:0,5% of reading ±1 digit
- Kalibrierung:pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
- Cat. No.:665-0277
- Anschlüsse:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe)
- Modell:A325
- Genauigkeit der Leitfähigkeit:±0,5% of reading ±1 digit
- Kalibrierung der Leitfähigkeit:Up to 5 points
- Sprachen:English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
- Schutzklasse:IP 67
- Kapazität Datenprotokollierung:5000 with date and time stamp
- B×T×H:240×100×65 mm
- pH - Auflösung:0,1 / 0,01 / 0,001
- Alarm:High/low limit alarm, calibration due alarm
- mV - Auflösung:0,1
- Batterielebensdauer:800 h
- Stromversorgung:AC adapter or 4 AA batteries
- pH - Kalibrierung:Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- Salinität - Bereich:0,06 - 80,00 psu / 0,05 - 42.00 ppt
- Temperaturbereich:–5,0...+105,0 °C
- Zellkonstante Leitfähigkeit:0.001 - 199.9 cm⁻¹
- Display:Graphic LCD with backlight
Accessories
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- 662-1140Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cableThermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus metersEinloggen für Verfügbarkeit1 * 1 SET - SFr. 94.40
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-ARProtective armor for portable meters, incl. hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, conductivity and dissolved oxygen sensor holderEinloggen für Verfügbarkeit1 * 1 ST - SFr. 126.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIO1210005Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the fieldEinloggen für Verfügbarkeit1 * 1 ST - SFr. 199.00
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-ESCDOrion™ Star A Series portable meter conductivity and DO probe holder, can be attached to either side of the portable meter armor and holds 12/15 mm Ø conductivity probes and a dissolved oxygen probe when placed in its calibration sleeveEinloggen für Verfügbarkeit1 * 1 ST - SFr. 36.50
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- 662-1547Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 89.50
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-CSHard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeveEinloggen für Verfügbarkeit1 * 1 ST - SFr. 178.00
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- ORIO1010053RS232 cable, for connecting a meter to a PC or printerOrion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 56.10
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-ESPHOrion™ Star™ A Series portable meter pH electrode holder, can be attached to the portable meter armor and holds one pH electrode when placed in the pH electrode storage sleeveEinloggen für Verfügbarkeit1 * 1 ST - SFr. 36.50
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- ORIO1010005Cable adapter, for connecting the RS232 port of a meter to a USB portOrion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 162.00



