Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215
Product Details & Documents
This benchtop meter is ideal for a wide range of applications and advanced pH and conductivity analysis in the lab. It offers two measuring channels allowing users to measure pH and conductivity simultaneously or view each channel separately.
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores ten most recent calibrations
- Non volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.
Measure pH, mV, relative mV or ORP with temperature on channel one and measure conductivity, TDS, salinity or resistivity with temperature on channel two.
Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values. Perform up to a five point conductivity calibration. Calibration editing option allows individual points to be fixed without a full recalibration. Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate.
Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures.
Certifications: CE, TUV 3-in-1 and FCC Class A certifications.
Delivery information: Supplied with electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable, meter test certificate and 3-year replacement warranty.
The kit additionally includes: 8157BNUMD ROSS Ultra Triode refillable epoxy-body pH/ATC electrode, 013005MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor, ROSS pH buffer and storage solution kit and Orion 1413 µS conductivity standard (5× 60 ml).
Technische Daten
- Bezeichnung:Bench pH/conductivity meter Orion Star A215, instrument alone
- Kalibrierpunkte:1 point relative mV (RmV) mode, 1 point ORP mode
- Auflösung der Leitfähigkeit:0,001 μS minimum, auto ranging, up to 4 significant digits
- Eingang:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
- Referenztemperatur:5 / 10 / 15 / 20 / 25 °C
- Anzahl der Kanäle:2
- TDS - Auflösung:0,001 ppm minimum, auto ranging, up to 4 significant digits
- Salinität - Genauigkeit:0,5% of reading ±1 digit
- Leitfähigkeitsbereich:0,001 µS/cm - 3000 mS/cm
- TDS - Bereich:0,001 - 200,0 ppm
- Temperatur - Kompensation:Linear, non linear nLFn, non linear nLFu, EP (USP), off
- mV - Bereich:±2000,0
- Resistivity resolution:1 ohm or 0,1 meg-ohm, auto ranging
- Spezifischer Widerstand - Bereich:2 Ω/cm - 100 MΩ/cm
- mV - Genauigkeit:±0,2 mV or ±0,05% of reading, whichever is greater
- Datenprotokollierung - Logfunktion:Manual / Automatic with AUTO-READ™
- Spezifischer Widerstand - Genauigkeit:0,5% of reading ±1 digit
- Operating temperature range:5...45 °C (ambient)
- Ausgänge:RS232, USB
- Temperaturgenauigkeit:±0,1 °C
- Temperaturauflösung:0,1 °C
- Konformität:IP 54
- Gewicht:0,9 kg
- pH - Genauigkeit:±0,002
- pH - Bereich:–2,000...+20,000
- Salinität - Auflösung:0,01 psu or 0,01 ppt minimum, auto ranging
- TDS - Genauigkeit:0,5% of reading ±1 digit
- Cat. No.:665-0271
- Kalibrierung:pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option
- Anschlüsse:BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
- Sensortyp:DuraProbe 4-cell conductivity sensor
- Modell:A215
- Genauigkeit der Leitfähigkeit:0,5% of reading ±1 digit > 3 μS, 0,5% of reading ±0,01 μS ≤ 3 μS
- Kalibrierung der Leitfähigkeit:Up to 5 points
- Schutzklasse:IP 54
- Kapazität Datenprotokollierung:2000 with date and time stamp
- B×T×H:240×180×110 mm
- pH - Auflösung:0,1 / 0,01 / 0,001
- Alarm:High/low limit alarm, calibration due alarm
- mV - Auflösung:0,1
- Batterielebensdauer:800 h
- Stromversorgung:AC adapter or 4 AA batteries
- pH - Kalibrierung:Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
- Salinität - Bereich:0,06 - 80,00 psu / 0,05 - 42,00 ppt
- Temperaturbereich:–5,0...+105,0 °C
- Zellkonstante Leitfähigkeit:0,001 - 199,9 cm⁻¹
- Display:Graphic LCD with backlight
Accessories
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- 662-1140Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cableThermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus metersEinloggen für Verfügbarkeit1 * 1 SET - SFr. 94.40
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- 662-1547Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 89.50
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-HBOrion™ Star™ beschwerte Basis der A-Serie zur Verwendung mit einem am Messgerät befestigten ElektrodenstativEinloggen für Verfügbarkeit1 * 1 ST - SFr. 69.60
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- ORIO1010005Cable adapter, for connecting the RS232 port of a meter to a USB portOrion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 162.00
Produktinformationen
- Katalognr.BezeichnungFürVerfügbarkeitPreis
- ORIO1010053RS232 cable, for connecting a meter to a PC or printerOrion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star metersEinloggen für Verfügbarkeit1 * 1 ST - SFr. 56.10
Produktinformationen
- Katalognr.BezeichnungVerfügbarkeitPreis
- ORIOSTARA-BEAAuf dem Tisch-Messgerät befestigtes Elektrodenstativ (Arm und Kopf der Elektrodenhalterung), Orion™ Star A-Serie, 360°-Bewegung (ohne beschwerte Basis)Einloggen für Verfügbarkeit1 * 1 ST - SFr. 202.00
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



