Heating tapes, HBS
Heating tape HBS, 5 m length, 1250 W
Catalog # HORS020307
Supplier: HORST
Jump to:
Product Details & Documents
The heating tapes are smooth, flexible and have a multiple glass-yarn insulation as well as an additional protection braid around the heating conductors.
Specifications
- Bezeichnung:Heating tape HBS, 5 m length, 1250 W
- Seq:07