Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215

Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215

Bench pH/conductivity meter Orion Star A215, kit incl. ROSS Ultra Triode pH/ATC probe and DuraProbe conductivity cell
Katalognr. 665-0272
Anbieter:   Thermo Orion
Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215
Katalognr. 665-0272
Anbieter:  Thermo Orion
Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215
Multi-parameter meter (pH/mV/conductivity), bench, Orion Star™ A215

Bench pH/conductivity meter Orion Star A215, kit incl. ROSS Ultra Triode pH/ATC probe and DuraProbe conductivity cell

Preis
SFr. 2’870.00
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

This benchtop meter is ideal for a wide range of applications and advanced pH and conductivity analysis in the lab. It offers two measuring channels allowing users to measure pH and conductivity simultaneously or view each channel separately.

  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Measurement stability indicator and selectable read modes – Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
  • Calibration log stores ten most recent calibrations
  • Non volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with universal power adapter or optional AA batteries

IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.

Measure pH, mV, relative mV or ORP with temperature on channel one and measure conductivity, TDS, salinity or resistivity with temperature on channel two.

Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values. Perform up to a five point conductivity calibration. Calibration editing option allows individual points to be fixed without a full recalibration. Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate.

Use linear, non linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures.

Certifications: CE, TUV 3-in-1 and FCC Class A certifications.

Informazioni per l'ordine: Supplied with electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable, meter test certificate and 3-year replacement warranty.
The kit additionally includes: 8157BNUMD ROSS Ultra Triode refillable epoxy-body pH/ATC electrode, 013005MD DuraProbe 4-cell (K=0,475) epoxy-body conductivity sensor, ROSS pH buffer and storage solution kit and Orion 1413 µS conductivity standard (5× 60 ml).

Technische Daten
  • Bezeichnung:
    Bench pH/conductivity meter Orion Star A215, kit incl. ROSS Ultra Triode pH/ATC probe and DuraProbe conductivity cell
  • Kalibrierpunkte:
    1 point relative mV (RmV) mode, 1 point ORP mode
  • Auflösung der Leitfähigkeit:
    0,001 μS minimum, auto ranging, up to 4 significant digits
  • Eingang:
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
  • Referenztemperatur:
    5 / 10 / 15 / 20 / 25 °C
  • Anzahl der Kanäle:
    2
  • TDS - Auflösung:
    0,001 ppm minimum, auto ranging, up to 4 significant digits
  • Salinität - Genauigkeit:
    0,5% of reading ±1 digit
  • Leitfähigkeitsbereich:
    0,001 µS/cm - 3000 mS/cm
  • TDS - Bereich:
    0,001 - 200,0 ppm
  • Temperatur - Kompensation:
    Linear, non linear nLFn, non linear nLFu, EP (USP), off
  • mV - Bereich:
    ±2000,0
  • Resistivity resolution:
    1 ohm or 0,1 meg-ohm, auto ranging
  • Spezifischer Widerstand - Bereich:
    2 Ω/cm - 100 MΩ/cm
  • mV - Genauigkeit:
    ±0,2 mV or ±0,05% of reading, whichever is greater
  • Datenprotokollierung - Logfunktion:
    Manual / Automatic with AUTO-READ™
  • Spezifischer Widerstand - Genauigkeit:
    0,5% of reading ±1 digit
  • Operating temperature range:
    5...45 °C (ambient)
  • Ausgänge:
    RS232, USB
  • Temperaturgenauigkeit:
    ±0,1 °C
  • Temperaturauflösung:
    0,1 °C
  • Konformität:
    IP 54
  • Gewicht:
    0,9 kg
  • pH - Genauigkeit:
    ±0,002
  • pH - Bereich:
    –2,000...+20,000
  • Salinität - Auflösung:
    0,01 psu or 0,01 ppt minimum, auto ranging
  • TDS - Genauigkeit:
    0,5% of reading ±1 digit
  • Kalibrierung:
    pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option
  • Cat. No.:
    665-0272
  • Anschlüsse:
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe), stirrer input (stirrer probe)
  • Sensortyp:
    DuraProbe 4-cell conductivity sensor
  • Modell:
    A215
  • Genauigkeit der Leitfähigkeit:
    0,5% of reading ±1 digit > 3 μS, 0,5% of reading ±0,01 μS ≤ 3 μS
  • Kalibrierung der Leitfähigkeit:
    Up to 5 points
  • Schutzklasse:
    IP 54
  • Kapazität Datenprotokollierung:
    2000 with date and time stamp
  • B×T×H:
    240×180×110 mm
  • pH - Auflösung:
    0,1 / 0,01 / 0,001
  • Alarm:
    High/low limit alarm, calibration due alarm
  • mV - Auflösung:
    0,1
  • Batterielebensdauer:
    800 h
  • Stromversorgung:
    AC adapter or 4 AA batteries
  • pH - Kalibrierung:
    Up to 5 points; Automatic buffer recognition (USA/NIST and DIN)
  • Salinität - Bereich:
    0,06 - 80,00 psu / 0,05 - 42,00 ppt
  • Temperaturbereich:
    –5,0...+105,0 °C
  • Zellkonstante Leitfähigkeit:
    0,001 - 199,9 cm⁻¹
  • Display:
    Graphic LCD with backlight
Accessories

Produktinformationen

  • Katalognr.
    Bezeichnung
    Für
    Verfügbarkeit
    Preis
  • 662-1140
    Printer and computer cable set, incl. one RS232 printer interface cable and one RS232 computer interface cable
    Thermo Scientific™ Orion™ Versa Star meters, Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters
    Einloggen für Verfügbarkeit
    1 * 1 SET - SFr. 94.40

Produktinformationen

  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • 662-1547
    Spare universal power adapter for Thermo Scientific™ Orion™ Star A series benchtop and portable meters and legacy Orion™ Star and Star Plus series meters
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 89.50

Produktinformationen

  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • ORIOSTARA-HB
    Orion™ Star™ beschwerte Basis der A-Serie zur Verwendung mit einem am Messgerät befestigten Elektrodenstativ
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 69.60

Produktinformationen

  • Katalognr.
    Bezeichnung
    Für
    Verfügbarkeit
    Preis
  • ORIO1010005
    Cable adapter, for connecting the RS232 port of a meter to a USB port
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series) and Orion™ Star and Star Plus meters
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 162.00

Produktinformationen

  • Katalognr.
    Bezeichnung
    Für
    Verfügbarkeit
    Preis
  • ORIO1010053
    RS232 cable, for connecting a meter to a PC or printer
    Orion™ Versa Star, Orion™ Star A series (A320, A220 + A210 series), Orion™ Dual Star benchtop meters, Orion 3-Star, 4-Star and 5-Star meters
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 56.10

Produktinformationen

  • Katalognr.
    Bezeichnung
    Verfügbarkeit
    Preis
  • ORIOSTARA-BEA
    Auf dem Tisch-Messgerät befestigtes Elektrodenstativ (Arm und Kopf der Elektrodenhalterung), Orion™ Star A-Serie, 360°-Bewegung (ohne beschwerte Basis)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 202.00

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..