Ac-DEVD-CHO
5 mg
Katalognr. 10404.
Anbieter: Biotium
Direkt zu
Product Details & Documents
Ac-DEVD-CHO is a synthetic tetrapeptide competitive inhibitor for Caspase-3/7, and contains the amino acid sequence of the PARP cleavage site.
- Soluble in DMSO
- Store at −20 °C
Ac-DEVD-CHO can be used to inhibit Caspase-3/7 activity in apoptotic cells, and to study events downstream of Caspase-3/7 activation.
Technische Daten
Technische Daten
- VE:5 mg
- Cat. No.:10404.
- Protein/Peptid-Name:Ac-DEVD-CHO
- Proteinsynonym:Caspase-3 inhibitor



